Mettler Toledo DO308 Single Sample Karl Fischer Drying Oven
Representative image only.
Mettler Toledo DO308 Single Sample Karl Fischer Drying Oven
Mettler Toledo | Mfr: 51371200| Item: 2575458
Enhance the versatility of your Mettler Toledo titrators
₹3,690,000.00INR / Each
₹3,690,000.00INR / Each
Buy It Together
Current Item
Mettler Toledo DO308  Single Sample Karl Fischer Drying Oven
Mettler Toledo DO308 Single Sample Karl Fischer Drying Oven
2575458
₹3,690,000.00INR / Each
+ + =
Total Price 9315000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Safe and easy water release
  • Simple and quick sample handling
More About this Item
Choose from a wide range of accessories compatible with your Mettler Toledo titrators for optimal performance.
The drying oven is ideal for accurate water determination of solid or viscous samples. Transferring samples into the heated zone is enabled by a simple transport system with a magnetic guide rod attached to the glass sample boat. The cold and the hot zone can be purged separately, so ambient moisture is quickly eliminated to reach a stable drift.
Specifications
  • DescriptionSingle Sample Karl Fischer Drying Oven