Mettler Toledo Gx0S Series Compact Potentiometric Titrators
Robust titrators with easy-to-use interface takes up little bench space
  • Compact yet performs a wide variety of potentiometric titrations
  • Easy-to-understand One Click® touchscreen user interface
  • Over 20 standard test methods preprogrammed
  • Automatic titrant and sensor recognition minimizes errors
  • Solvent manager provides safe chemical handling
  • Flexible data export via two USB ports
Use these compact potentiometric titrators to perform routine quantitative chemical analysis with high accuracy, flexibility, and reproducibility. They are useful for acid/base titration, chromatometric titration, redox titration, precipitation titration, voltametric titration, and more. Robust design and easy-to-use interface make these simple to use.
Use the large color touchscreen to operate the simple-to-use One Click® user interface to customize home screens to run your workflows with a single tap. Each user can set their own home screen with customized language selection (English, German, French, Spanish, Italian, Chinese, Russian, Polish, Korean, or Portuguese) and shortcut keys.
Automatic titrant and sensor recognition reduces extra data entry steps and helps minimize errors. Ensure user safety with the solvent manager. It is fully controlled by the titrator, and offers filling, draining, and solvent exchange without additional user interaction. For secure titrator operation, designate user groups to allow full or limited access. Easily export data in CSV, PDF, or XML files using a memory stick or connect to a printer via the USB port.
4,340,000.00 - ₹8,542,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Output Power (VAC) Description
Availability Pricing
Mettler Toledo G10S Compact Potentiometric Titrator, 5 Analysis Methods 7890723 Mfr # 30267117
COMPARE
USB (2), Ethernet 100 to 240 G10S Compact Potentiometric Titrator, 5 Analysis Methods
₹4,340,000.00INR / Each
Mettler Toledo G20S Compact Potentiometric Titrator, 150 Analysis Methods 7890724 Mfr # 30252669
COMPARE
USB (2), Ethernet 100 to 240 G20S Compact Potentiometric Titrator, 150 Analysis Methods
₹6,301,000.00INR / Each
Mettler Toledo G20S  Rondolino Automated Compact Potentiometric Titrator 7890725 Mfr # 30252671
COMPARE
100 to 240 G20S Rondolino Automated Compact Potentiometric Titrator
₹8,542,000.00INR / Each