Mettler Toledo DH100 Heating System; 110 V

Representative image only.

Mettler Toledo DH100 Heating System; 110 V

Mettler Toledo | Mfr: 51108779| Item: 2575493

Enhance the versatility of your Mettler Toledo titrators

₹2,339,000.00INR / Each

₹2,339,000.00INR / Each

Buy It Together

Current Item

Mettler Toledo DH100 Heating System; 110 V
Mettler Toledo DH100 Heating System; 110 V
2575493

₹2,339,000.00INR / Each
+ + =
Total Price 7964000.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Choose from a wide range of accessories compatible with your Mettler Toledo titrators for optimal performance.

The DH100 heating system facilitates dissolution or allows chemical reactions to complete during preparation and analysis when elevated temperatures are required. The system allows both controlled heating as well as temperature control during titration up to a maximum temperature of 100 °C.

Specifications
  • DescriptionHeating System; 110 V
Reviews
Q&A