Mettler Toledo Compact Stirrer
Representative image only.
Mettler Toledo Compact Stirrer
Mettler Toledo | Mfr: 51109150| Item: 2575381
Enhance the versatility of your Mettler Toledo titrators
₹510,000.00INR / Each
₹510,000.00INR / Each
Buy It Together
Current Item
Mettler Toledo Compact stirrer
Mettler Toledo Compact stirrer
2575381
₹510,000.00INR / Each
+ + =
Total Price 6135000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Two impeller stirrers.
More About this Item
Choose from a wide range of accessories compatible with your Mettler Toledo titrators for optimal performance.
This compact stirrer provides efficient stirring whenever magnetic stirring is impractical. Offers higher stirring speed and strength than built in magnetic stirrers for titration and/or pH measurement.
Specifications
  • DescriptionCompact Stirrer