Mettler Toledo Bottle Adapter, 40.5 mm dia, 3.5 mm Slope, Merck

Representative image only.

Mettler Toledo Bottle Adapter, 40.5 mm dia, 3.5 mm Slope, Merck

Mettler Toledo | Mfr: 23774| Item: 2575683

Enhance the versatility of your Mettler Toledo titrators

₹31,600.00INR / Each

₹31,600.00INR / Each

Buy It Together

Current Item

Mettler Toledo Bottle Adapter, 40.5 mm dia, 3.5 mm Slope, Merck
Mettler Toledo Bottle Adapter, 40.5 mm dia, 3.5 mm Slope, Merck
2575683

₹31,600.00INR / Each
+ + =
Total Price 5656600.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Choose from a wide range of accessories compatible with your Mettler Toledo titrators for optimal performance.

Specifications
  • DescriptionBottle Adapter, 40.5 mm dia, 3.5 mm Slope, Merck
Reviews
Q&A