Mettler Toledo Bottle Adapter, 37.5 mm dia, 5 mm Slope, USA

Representative image only.

Mettler Toledo Bottle Adapter, 37.5 mm dia, 5 mm Slope, USA

Mettler Toledo | Mfr: 23787| Item: 2575684

Enhance the versatility of your Mettler Toledo titrators

₹24,200.00INR / Each

₹24,200.00INR / Each

Buy It Together

Current Item

Mettler Toledo Bottle Adapter, 37.5 mm dia, 5 mm Slope, USA
Mettler Toledo Bottle Adapter, 37.5 mm dia, 5 mm Slope, USA
2575684

₹24,200.00INR / Each
+ + =
Total Price 5649200.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Choose from a wide range of accessories compatible with your Mettler Toledo titrators for optimal performance.

Specifications
  • DescriptionBottle Adapter, 37.5 mm dia, 5 mm Slope, USA
Reviews
Q&A