Mark-10 AC1030 AC adapter/charger, 110V US

Representative image only.

Mark-10 AC1030 AC adapter/charger, 110V US

Mark-10 | Mfr: AC1030| Item: 2530280

₹12,700.00INR / Each

₹12,700.00INR / Each

Buy It Together

Current Item

Mark-10 AC1030 AC adapter/charger, 110V US
Mark-10 AC1030 AC adapter/charger, 110V US
2530280

₹12,700.00INR / Each
+ =
Total Price 482700.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • AC1030 replaces 08-1007
  • 3 year warranty
  • 3 year warranty

More About this Item

Expand the testing capabilities of your Mark-10 force and torque systems with these parts and accessories.

Adapter is compatible with any Mark-10 force or torque gauge, except Series MG and MGT gauges manufactured before September, 2010.

Conformite Europeenne3 Year Warranty
Specifications
  • DescriptionAC Adapter for M2 Force Gauges, 110 VAC
  • Warranty3 year
Reviews
Q&A