MadgeTech PRTEMP1000-30-PSIG Temp/Pressure Recorder; 30 psig, 1/4"NPT
Representative image only.
MadgeTech PRTEMP1000-30-PSIG Temp/Pressure Recorder; 30 psig, 1/4"NPT
MadgeTech | Mfr: PRTEMP1000-30-PSIG| Item: 1800333
Call For Price
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
MadgeTech PRTEMP1000-30-PSIG Temp/Pressure Recorder; 30 psig, 1/4"NPT
MadgeTech PRTEMP1000-30-PSIG Temp/Pressure Recorder; 30 psig, 1/4"NPT
1800333
Call For Price
+ =
Total Price 44700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
The MH1000PR series is a battery powered, stand-alone pressure and temperature data logger. This all-in-one compact, portable, easy to use device measures and records up to 16,383 pressure and 16.383 temperature measurements. The data logger accommodates numerous adaptors for various pressure ranges and pipefittings. Its real time clock ensures that all data is time and date stamped. The storage medium is non-volatile solid-state memory, providing maximum data security even if the battery becomes discharged. The device can be started and stopped directly from your computer and its small size allows it to fit almost anywhere. Data retrieval is quick and easy. Simply plug a logger into an empty COM port and our user-friendly software does the rest.
FEATURES
  • Rugged, stainless steel case
  • Submersible
  • Programmable start time
  • Real-time operation
  • User-friendly
  • Low cost
  • ¼" NPT fitting
  • Readings selectable 1 every 2 sec to 1 every 12 hour
1 Year Warranty
Specifications
  • Number of Channels1
  • Input TypeTemperature and pressure
  • Max Temperature (° F)176
  • Min Temperature (° F)-40
  • Max Temperature (° C)80
  • Min Temperature (° C)-40
  • DescriptionTemp/Pressure Recorder; 30 psig, 1/4"NPT