Lascar Electronics EL-USB-LITE USB Temperature Data Logger, 4,000+ readings, 1 month battery life
Representative image only.
Lascar Electronics EL-USB-LITE USB Temperature Data Logger, 4,000+ readings, 1 month battery life
Lascar Electronics | Mfr: EL-USB-LITE| Item: 2303951
Direct-to-USB connection and easy-to-use functionality
Call For Price
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Lascar Electronics EL-USB-LITE USB Temperature Data Logger, 4,000+ readings, 1 month battery life
Lascar Electronics EL-USB-LITE USB Temperature Data Logger, 4,000+ readings, 1 month battery life
2303951
Call For Price
+ =
Total Price 44700.00 ADD 2 ITEMS TO CART
Product Description
Includes
configuration software and battery.
Key Features
  • User-programmable alarm thresholds — perfect for compliance monitoring
  • All data loggers come with software
  • 1 year warranty
More About this Item
Simply insert any of these compact, 4.5" (11.4 cm) long loggers into your computer USB port, name it, select high and low alarms if required, and set the logger start time. Once configured, the logger can be removed from the computer to log independently. Data is downloaded back onto the PC where it can be analyzed and viewed as a graph using included software.
The low-cost model EL-USB-LITE stores over 4000 readings and has a fixed 30-minute sampling rate and a measurement range of 14 to 122°F (–10 to 50°C).
1 Year Warranty
Specifications
  • Number of Channels1
  • Input TypeTemperature (one internal)
  • Max Temperature (° F)122
  • Min Temperature (° F)14
  • Max Temperature (° C)50
  • Min Temperature (° C)-10
  • Temperature Accuracy (° F)±4.5
  • Temperature Accuracy (° C)±2.5
  • Sampling Rate1 per 30 minutes
  • Memory Size4,000+ data points
  • Battery Life1 month
  • Battery Type1 CR1620 (included)
  • Display TypeNo
  • DescriptionUSB Temperature Data Logger, 4,000+ readings, 1 month battery life