Lascar Electronics EL-USB-CO300 Carbon monoxide Data Logger, 0 to 300 ppm, 32,000+ readings, 3 month battery life
Representative image only.
Lascar Electronics EL-USB-CO300 Carbon monoxide Data Logger, 0 to 300 ppm, 32,000+ readings, 3 month battery life
Lascar Electronics | Mfr: EL-USB-CO300| Item: 2303968
Direct-to-USB connection and easy-to-use functionality
Call For Price
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Lascar Electronics EL-USB-CO300 Carbon monoxide Data Logger, 0 to 300 ppm, 32,000+ readings, 3 month battery life
Lascar Electronics EL-USB-CO300 Carbon monoxide Data Logger, 0 to 300 ppm, 32,000+ readings, 3 month battery life
2303968
Call For Price
+ =
Total Price 44700.00 ADD 2 ITEMS TO CART
Product Description
Includes
configuration software and battery.
Key Features
  • User-programmable alarm thresholds
  • All data loggers come with software
  • 1 year warranty
More About this Item
Simply insert any of these compact, 4.5" (11.4 cm) L loggers into your computer USB port, name it, choose your required sampling rate, select high and low alarms if required, and set the logger start time. Once configured, the logger can be removed from the computer to log independently. Data is downloaded back onto the PC where it can be analyzed and viewed as a graph using included software.
Use model 23039-58 for a 0 to 1000 ppm range and model 23039-68 for a 0 to 300 ppm measurement range; both store more than 32,000 readings. An bright red LED will flash and an audible alarm will sound when the user-determined level is exceeded.
1 Year WarrantyConformite Europeenne
Specifications
  • Number of Channels1
  • Input TypeCarbon monoxide (0 to 300 ppm; 0.5 ppm CO resolution; ±4 ppm CO accuracy)
  • Memory Size32,510
  • Max Operating Temperature (° F)104
  • Min Operating Temperature (° F)14
  • Battery Life3 months (typical)
  • Display TypeNo
  • DescriptionCarbon monoxide Data Logger, 0 to 300 ppm, 32,000+ readings, 3 month battery life