Lascar Electronics EL-USB-1-PRO High Temperature Data Logger, 32,000+ readings, 3 year battery life
Representative image only.
Lascar Electronics EL-USB-1-PRO High Temperature Data Logger, 32,000+ readings, 3 year battery life
Lascar Electronics | Mfr: EL-USB-1-PRO| Item: 2303966
Direct-to-USB connection and easy-to-use functionality
Call For Price
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Lascar Electronics EL-USB-1-PRO High Temperature Data Logger, 32,000+ readings, 3 year battery life
Lascar Electronics EL-USB-1-PRO High Temperature Data Logger, 32,000+ readings, 3 year battery life
2303966
Call For Price
+ =
Total Price 44700.00 ADD 2 ITEMS TO CART
Product Description
Includes
configuration software and battery.
Key Features
  • User-programmable alarm thresholds — perfect for compliance monitoring
  • All data loggers come with software
  • 1 year warranty
More About this Item
Simply insert any of these compact, 4.5" (11.4 cm) long loggers into your computer USB port, name it, select high and low alarms if required, and set the logger start time. Once configured, the logger can be removed from the computer to log independently. Data is downloaded back onto the PC where it can be analyzed and viewed as a graph using included software.
For application requiring an extended temperature range, model EL-USB-1-PRO measures from –40 to 257°F (–40 to 125°C), stores over 32,000 readings, and includes a waterproof stainless steel enclosure for more hostile environments. While in SS case, unit may be used in an autoclave.
1 Year WarrantyConformite EuropeenneIP 67
Specifications
  • Number of Channels1
  • Input TypeTemperature (one internal)
  • Max Temperature (° F)257
  • Min Temperature (° F)-40
  • Min Temperature (° C)-40
  • Max Temperature (° C)125
  • Temperature Resolution (° F)0.2
  • Temperature Accuracy (° F)±0.4
  • Temperature Resolution (° C)0.1
  • Temperature Accuracy (° C)±0.2
  • Sampling Rate1 per 1 second to 1 per 12 hours
  • Memory Size32,510 data points
  • Battery Life2 years (typical)
  • Battery Type1 lithium 2/3 AA (included)
  • Display TypeNo
  • Length (in)4 21/64
  • Width (in)11/16
  • DescriptionHigh Temperature Data Logger, 32,000+ readings, 3 year battery life