Lascar Electronics EL-USB-1-LCD USB Temperature Data Logger with Display
Representative image only.
Lascar Electronics EL-USB-1-LCD USB Temperature Data Logger with Display
Lascar Electronics | Mfr: EL-USB-1-LCD| Item: 2303953
Direct-to-USB connection and easy-to-use functionality
Call For Price
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Lascar Electronics EL-USB-1-LCD USB Temperature Data Logger with Display
Lascar Electronics EL-USB-1-LCD USB Temperature Data Logger with Display
2303953
Call For Price
+ =
Total Price 44700.00 ADD 2 ITEMS TO CART
Product Description
Includes
configuration software and battery.
Key Features
  • User-programmable alarm thresholds — perfect for compliance monitoring
  • All data loggers come with software
  • USB interface for set-up and data download
  • User-programmable alarm thresholds
  • Status indication via red, green and orange LEDs
  • High contrast LCD, with two and a half digit temperature display function
  • Immediate, delayed, and push-to-start logging options
  • 1 year warranty
More About this Item
Simply insert any of these compact, 4.5" (11.4 cm) long loggers into your computer USB port, name it, select high and low alarms if required, and set the logger start time. Once configured, the logger can be removed from the computer to log independently. Data is downloaded back onto the PC where it can be analyzed and viewed as a graph using included software.
For ambient temperature measurements between –31 to 176°F (–35 to 80°C), model EL-USB-1-LCD stores over 16,000 readings. Much like the EL-USB-1, but with an LCD display for quick check of the current, minimum, or maximum temperature recorded.
The data logger is protected against moisture to IP67 standard when the protective cap is fitted.
1 Year Warranty
Specifications
  • Number of Channels1
  • Input TypeTemperature (one internal)
  • Max Temperature (° F)176
  • Min Temperature (° F)-31
  • Max Temperature (° C)80
  • Min Temperature (° C)-35
  • Temperature Resolution (° F)1
  • Temperature Accuracy (° F)±2
  • Temperature Resolution (° C)0.5
  • Temperature Accuracy (° C)±1
  • Sampling Rate1 per 10 seconds to 1 per 12 hours
  • Memory Size16,382 data points
  • Battery Life2 years (typical)
  • Battery Type1 lithium 1/2 AA (included)
  • Display TypeYes
  • Length (in)4 45/64
  • Width (in)1
  • DescriptionUSB Temperature Data Logger with Display