Labconco LabSolutions™ Neutralizing Acid Rinse; 1 L
Representative image only.
Labconco LabSolutions™ Neutralizing Acid Rinse; 1 L
Labconco | Mfr: 4522200| Item: 9896076
Removes wide range of lab contaminants
Call For Price
Call For Price
Buy It Together
Current Item
Labconco LabSolutions™ Neutralizing Acid Rinse; 1 L
Labconco LabSolutions™ Neutralizing Acid Rinse; 1 L
9896076
Call For Price
+ =
Total Price 9300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Rinses completely
  • Corrects pH, reducing alkaline detergent residue
  • Removes hard water deposits
More About this Item
LabSolutions detergents are specially formulated for Labconco glassware washers.
Neutralizing acid rinse provides pH correction due to alkaline detergents. Easily and safely removes hard water, mineral, and lime scale deposits. Formulated for Labconco FlaskScrubber and SteamScrubber washers.
Specifications
  • Capacity (Liters)1
  • Capacity (Gal)0.26
  • MaterialMild acid
  • Compatible WithAutomatic glassware washers
  • DescriptionNeutralizing Acid Rinse