Labconco FreeZone Heated Product Shelf Freeze Drying Chamber, 5 Shelves, with Valves, 230V, EU, CN/AU, UK, & India Plugs
Representative image only.
Labconco FreeZone Heated Product Shelf Freeze Drying Chamber, 5 Shelves, with Valves, 230V, EU, CN/AU, UK, & India Plugs
Labconco | Mfr: 780802190| Item: 0334619
Easily attach flasks to your freeze dry system or contain samples in bulk
₹4,413,000.00INR / Each
₹4,413,000.00INR / Each
Buy It Together
Current Item
Labconco FreeZone Heated Product Shelf Freeze Drying Chamber, 5 Shelves, with Valves, 230V, EU, CN/AU, UK, & India Plugs
Labconco FreeZone Heated Product Shelf Freeze Drying Chamber, 5 Shelves, with Valves, 230V, EU, CN/AU, UK, & India Plugs
0334619
₹4,413,000.00INR / Each
+ + + =
Total Price 4494800.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Acrylic cylinder and base, temperature probes, five removeable shelves, and EU, CN/AU, UK, & India plugs.
Key Features
  • Easily mounts atop FreeZone Freeze Dry Systems
  • Accommodates ½” and ¾” flask adapters
  • Compatible with Labconco freeze dryers
  • 8 month warranty
More About this Item
These drying chambers provide a means to attach flasks to your freeze dryer or contain samples in bulk. Various models are available from 8-, 12- to 16-port drying chambers to 3 to 5 heated product shelf chambers.
The 12" dia acrylic drying chamber features 8 ports that accommodate both ½” and ¾” adapters for quick and easy connection of flasks. The five 10" dia removable shelves are are intended to be used in conjunction with Labconco Freeze Dryers to facilitate the lyophilization process. Each shelf is equipped with a shelf sensor to accurately control the temperature of the shelf. .
Conformite Europeenne
Specifications
  • Cooling Capacity (° F)140
  • Cooling Capacity (° C)50
  • Exterior Width (InchShort)13.5
  • Exterior Height (InchShort)24.6
  • Exterior Depth (in)15.2
  • Power (VAC)230
  • Power (Hz)50
  • DescriptionHeated Product Shelf Freeze Drying Chamber, 5 Shelves, with Valves, 230V, EU, CN/AU, UK, & India Plugs
  • Warranty8 Months