Labconco FreeZone Freeze Drying Manifold, 20 Ports; FreeZone FD

Representative image only.

Labconco FreeZone Freeze Drying Manifold, 20 Ports; FreeZone FD

Labconco | Mfr: 7522300| Item: 0334831

Manifolds are ideal for freeze-drying contents of serum vials, ampules, or small flasks

Call For Price

Call For Price

Buy It Together

Current Item

Labconco FreeZone Freeze Drying Manifold, 20 Ports; FreeZone FD
Labconco FreeZone Freeze Drying Manifold, 20 Ports; FreeZone FD
0334831

Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Easily mounts atop FreeZone Freeze Dry Systems
  • Accommodates ½” and ¾” adapters for quick connection of flasks
  • Compatible with 1-, 2.5-, 4.5-, 6-, 12-, and 18-L FreeZone Freeze Dryers
  • 1 year warranty

More About this Item

Stainless steel manifolds are ideal for freeze-drying contents of serum vials, ampules, or small flasks.

1 Year Warranty
Specifications
  • Capacity (Liters)12
  • Cooling Capacity (° F)-58
  • Cooling Capacity (° C)-50
  • Sublimation Rate (Liters/day)8
  • Exterior Width (InchShort)31.875
  • Exterior Height (InchShort)36.5
  • Exterior Depth (in)28
  • Compressor Horse Power (hp)0.75
  • Power (VAC)220
  • Power (Hz)50
  • DescriptionFreeze Drying Manifold, 20 Ports; FreeZone FD
  • Warranty1 Year
Reviews
Q&A