Labconco Fast-Freeze™ 7541101 Amber Glass Drying Flask; 750 mL
Representative image only.
Labconco Fast-Freeze™ 7541101 Amber Glass Drying Flask; 750 mL
Labconco | Mfr: 7541101| Item: 0335334
Provides easy and efficient handling of freeze dry samples
₹109,000.00INR / Each
₹109,000.00INR / Each
Buy It Together
Current Item
Labconco Fast-Freeze™ 7541101 Amber Glass Drying Flask; 750 mL
Labconco Fast-Freeze™ 7541101 Amber Glass Drying Flask; 750 mL
0335334
₹109,000.00INR / Each
+ + + + + +
Labconco FreeZone Fast-Freeze Flask, Amber, Glass, 750 mL
0335330
₹69,900.00INR / Each
=
Total Price 1067900.00 ADD 7 ITEMS TO CART
Product Description
Includes
silicone rubber top and supply of filter paper.
Key Features
  • Borosilicate glass withstands extreme temperatures and high vacuum
  • Amber-colored glass protects light-sensitive samples
  • Compatible with any brand freeze dryer with ¾” diameter valve ports
  • Flat bottom and wide mouth design provides easy loading and cleaning
More About this Item
The Fast-Freeze™ drying flask is constructed of high strength borosilicate glass which can withstand a maximum temperature of 490º C. It easily attaches to the valve ports on Labconco drying chambers, manifolds, and freeze dryer systems with the use of an adapter (sold separately).
Specifications
  • Capacity (Liters)0.75
  • Height (InchShort)7 7/8
  • DescriptionAmber Glass Drying Flask; 750 mL