Labconco FreeZone 2.5-Liter Freeze Dryers
Designed for lyophilizing light sample loads in a limited floor space
  • Full color, 5" capacitive touchscreen
  • Moisture sensor prevents refrigeration or vacuum start-up when moisture in the drain line is detected
  • Vaccum break valve bleeds air into system when power is shut off
These FreeZone freeze dryers have an upright stainless steel or PTFE collector coil that holds 2.5 liters of ice before defrosting. Collector lid, 3/4" thick, has a neoprene gasket and 3" diameter port for connection of drying accessories (sold separately). The -50ºC (-58ºF) dryers have a single 1/3 hp HCFC/CFC-free refrigeration system which are suitable for aqueous samples, while the -84ºC (-119ºF) have two 1/3 hp systems and are suitable for samples with low eutectic points. The dryers are easy to operate due to the full color 5" touch screen control that comes compete with Lyo-Works™ OS. Real time display shows collector temperature and vacuum level. Setting include 5 different languages, time, time zone, temperature (ºC or ºF), and vacuum (mBar, Torr, or Pa). Other setting available include, alerts, display, sounds, screen time out, and maintenance alert.
Level-specific security lock-out prevents changes to programs and other system parameters. Data logging stores and displays data in table or chart format and can be exported via the side mounted USB port or rear mounted Ethernet connection. Other features include; Moisture sensor, vacuum control valve, vacuum break valve, side mounted quick-disconnect drain hose fittings, and rear mounted electrical receptacle for vacuum pump connection.
All models require (not included) vacuum pump with a displacement of at leat 98 liters per minute, 0.002 mBar ultimate pressure and fitting suitable for 3/4" ID vacuum hose, drying accessories, and freeze dry glassware.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cooling Capacity (° C) Power (VAC) Description
Availability Pricing
Labconco FreeZone Benchtop Freeze Dryer with PTFE Coil, 2.5L -84°C; 230V 60Hz North America 0333611 Mfr # 710202010
COMPARE
-84 230 Freeze Dryer with PTFE Coil, 230V 60Hz, North American Plug
Call For Price
Labconco FreeZone Benchtop Freeze Dryer with PTFE Coil, 2.5L -84°C; 230V 50Hz Schuko 0333613 Mfr # 710202030
COMPARE
-84 230 Freeze Dryer with PTFE Coil, 230V 50Hz, EU Plug