Julabo SW22 and 23 SW All-purpose Spring Tray for Flasks
Representative image only.
Julabo SW22 and 23 SW All-purpose Spring Tray for Flasks
Julabo | Mfr: 8970630| Item: 1214952
Quality accessories to add to your shaker bath
₹227,000.00INR / Each
₹227,000.00INR / Each
Buy It Together
Current Item
Julabo SW22 and 23 SW All-purpose Spring Tray for Flasks
Julabo SW22 and 23 SW All-purpose Spring Tray for Flasks
1214952
₹227,000.00INR / Each
+
Julabo SW22 Shaking Water Bath, ±0.2 C, 115V
1214900
Call For Price
=
Total Price 227000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from lids, racks and trays for your Julabo shaker
  • Includes springs for Erlenmeyer flask from 25 to 1000 mL size
  • Pre-assebled stainless steel tray
  • 2 year warranty
More About this Item
Allows flexibility of flask size on one platform. Fits Julabo SW22 and 23 Shaking Water Baths (12149-00, 12149-05, 12149-10, and 12149-15)
Two year warranty
Specifications
  • DescriptionSW All-purpose Spring Tray for Flasks