Julabo PURA™ Flat Bath Cover for PURA 22 Water Bath, 6 x 115 mm Diameter Openings
Representative image only.
Julabo PURA™ Flat Bath Cover for PURA 22 Water Bath, 6 x 115 mm Diameter Openings
Julabo | Mfr: 9970574| Item: 1215394
Optimize performance of your Julabo PURA™ water baths
₹227,000.00INR / Each
₹227,000.00INR / Each
Buy It Together
Current Item
Julabo PURA™ Flat Bath Cover for PURA 22 Water Bath, 6 x 115 mm Diameter Openings
Julabo PURA™ Flat Bath Cover for PURA 22 Water Bath, 6 x 115 mm Diameter Openings
1215394
₹227,000.00INR / Each
+ =
Total Price 581000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
flat bath cover and set of rings.
Key Features
  • Choose from covers, cooling coils, and more to enhance your water bath
  • Bath cover is suitable for use with PURA 22 water bath
  • 2 Year Parts & Labor Warranty
More About this Item
These Julabo PURA™ water bath accessories can enhance performance of your water bath. Choose from different bath covers, drain tap, cooling coils, and more. Water baths sold separately.
2 Year Warranty
Specifications
  • Compatible WithJulabo PURA™ 22 Water Bath
  • Diameter (mm)115
  • DescriptionFlat Bath Cover for PURA 22 Water Bath, 6 x 115 mm Diameter Openings
  • Warranty2 Year Parts & Labor