Julabo PURA™ Water Baths
Easy to use and easy to clean
  • Easily view bright white LED display from across the room
  • Built-in dry running protection
  • Meets Class I (NFL) according to DIN 12876-1
  • All wetted parts are constructed of stainless steel for optimal chemical resistance
These Julabo PURA™ water baths are designed with features that make everyday laboratory task easier, including durable side handles that allow the bath to be easily moved and easy-to-read LED display. These baths also feature an integrated timer and a specially designed outer edge, which automatically drains condensed fluid back into the bath. Other features include a removeable platform, non-slip rubber feet, and controls on the front of the unit for ease of operation. All models, except the PURA 4, come with an integrated drain screw for quick and clean draining of the baths.
354,000.00 - ₹597,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Immersion Depth (in) Power (VAC) Reservoir Capacity (L)
Availability Pricing
Julabo PURA™ 4 Water Bath, 4.5 L, 18 to 99.9 °C, 0.5 kW Heater; 115 VAC, 60 Hz 1215375 Mfr # 9550504.22
COMPARE
5 1/2 115 4.5
₹354,000.00INR / Each
Julabo PURA™ 10 Water Bath, 9.5 L, 18 to 99.9 °C, 1.2 kW Heater; 115 VAC, 60 Hz 1215376 Mfr # 9550510.22
COMPARE
5 1/2 115 9.5
₹423,000.00INR / Each
Julabo PURA™ 14 Water Bath, 14 L, 18 to 99.9 °C, 1.3 kW Heater; 115 VAC, 60 Hz 1215377 Mfr # 9550514.22
COMPARE
5 1/2 115 14
₹461,000.00INR / Each
Julabo PURA™ 22 Water Bath, 25.5 L, 18 to 99.9 °C, 1.3 kW Heater; 115 VAC, 60 Hz 1215378 Mfr # 9550522.22
COMPARE
7 1/10 115 25.5
₹506,000.00INR / Each
Julabo PURA™ 22 Water Bath, 25.5 L, 18 to 99.9 °C, 2 kW Heater; 230 VAC, 50/60 Hz 1215379 Mfr # 9550522.33
COMPARE
7 1/10 230 25.5
₹506,000.00INR / Each
Julabo PURA™ 30 Water Bath, 36 L, 18 to 99.9 °C, 1.3 kW Heater; 115 VAC, 60 Hz 1215380 Mfr # 9550530.22
COMPARE
7 1/10 115 36
₹597,000.00INR / Each
Julabo PURA™ 30 Water Bath, 36 L, 18 to 99.9 °C, 2 kW Heater; 230 VAC, 50/60 Hz 1215381 Mfr # 9550530.33
COMPARE
7 1/10 230 36
₹597,000.00INR / Each
Showing 1- 7 of 7