Julabo PURA™ Cooling Coil for PURA 10 and 14 Water Baths, Stainless Steel
Representative image only.
Julabo PURA™ Cooling Coil for PURA 10 and 14 Water Baths, Stainless Steel
Julabo | Mfr: 9970534| Item: 1215388
Optimize performance of your Julabo PURA™ water baths
₹137,000.00INR / Each
₹137,000.00INR / Each
Buy It Together
Current Item
Julabo PURA™ Cooling Coil for PURA 10 and 14 Water Baths, Stainless Steel
Julabo PURA™ Cooling Coil for PURA 10 and 14 Water Baths, Stainless Steel
1215388
₹137,000.00INR / Each
+ =
Total Price 491000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
cooling coil and 10.5mm hose barbs.
Key Features
  • Choose from covers, cooling coils, and more to enhance your water bath
  • Stainless steel cooling coil inserts beneath false bottom of PURA 10 and PURA 14 water baths
  • A hose with an inner diameter of 10/12 mm is required
  • 2 Year Parts & Labor Warranty
More About this Item
These Julabo PURA™ water bath accessories can enhance performance of your water bath. Choose from different bath covers, drain tap, cooling coils, and more. Water baths sold separately.
2 Year Warranty
Specifications
  • MaterialStainless Steel
  • Compatible WithJulabo PURA™ 10 and 14 Water Bath
  • DescriptionCooling Coil for PURA 10 and 14 Water Baths, Stainless Steel
  • Warranty2 Year Parts & Labor