InnoCal NIST-Traceable Calibration, Hart Temperature RTD Meter and Probe System
Representative image only.
InnoCal NIST-Traceable Calibration, Hart Temperature RTD Meter and Probe System
InnoCal | Item: 1700211
Regardless of the type of thermometry probe your application requires, InnoCal can ensure the accuracy of your meter, probe, or system
₹128,000.00INR / Each
₹128,000.00INR / Each
Buy It Together
Current Item
InnoCal NIST-Traceable Calibration, Hart Temperature RTD Meter and Probe System
InnoCal NIST-Traceable Calibration, Hart Temperature RTD Meter and Probe System
1700211
₹128,000.00INR / Each
+ +
Traceable Thermistor Mini Thermometer
0840260
₹6,400.00INR / Each
+ +
Traceable High-Precision RTD Thermometer
3780392
₹61,500.00INR / Each
+ +
Traceable Scientific Thermistor Thermometer with USB Probe
9008009
₹81,700.00INR / Each
+ +
Traceable Type K Thermocouple Thermometer with Memory
9121005
₹54,200.00INR / Each
+ =
Total Price 388100.00 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification
  • EPA, USDA, ISO and GLP/GMPs standards require regular NIST-traceable calibration of all test equipment being used in critical applications
  • Calibration service for a hart temperature RTD meter and probe system
  • 1 year warranty
More About this Item
Regardless of the type of thermometry probe your application requires, InnoCal can ensure the accuracy of your meter, probe, or system!
All measurements reported are traceable to SI units via national standards maintained by NIST and performed in compliance with ANSI Z540, ISO17025 and MIL-STD-45662A. See the sample certificate under the “More Info” section to see how InnoCal’s certificates provide more information in a concise manner than the rest of the industry.
InnoCal is accredited to ISO/IEC 17025 by the American Association for Laboratory Accreditation (A2LA) under certificate number 1746.01. This accreditation is your assurance that the calibration is performed in a qualified, controlled laboratory which is technically proficient and committed to quality and customer-focused continuous improvement. Our scope of A2LA accreditation can be found under the “More Info” section.
Calibrations on temperature systems (meter and probe) are performed using direct comparison in high-stability liquid using high-precision and SPRT’s and displays. Measurements can be performed at four points between -196 and 1300°C. Points are generally selected at standard ITS-90 points across the range of the probe but customer-specified points can be accommodated. Calibrating the probe and meter together produces the highest accuracies.
1 Year Warranty
Specifications
  • Calibration TypeTemperature
  • Instrument Only CalibrationYes
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points4 points
  • DescriptionNIST-Traceable Calibration; Hart Temperature RTD Meter and Probe System