InnoCal NIST-Traceable Calibration; Calibration Weight/Mass, 0.51 to 2 lb; ASTM 1-3; OIML F1/F2
Representative image only.
InnoCal NIST-Traceable Calibration; Calibration Weight/Mass, 0.51 to 2 lb; ASTM 1-3; OIML F1/F2
InnoCal | Item: 1710828
Mass Value Report
₹18,700.00INR / Each
₹18,700.00INR / Each
Buy It Together
Current Item
InnoCal NIST-Traceable Calibration; Calibration Weight/Mass, 0.51 to 2 lb; ASTM 1-3; OIML F1/F2
InnoCal NIST-Traceable Calibration; Calibration Weight/Mass, 0.51 to 2 lb; ASTM 1-3; OIML F1/F2
1710828
₹18,700.00INR / Each
+ =
Total Price 55900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Measurements are traceable to NIST
  • Certificate lists actual weight values, uncertainties, and tolerances
More About this Item
Mass calibrations are performed by an ISO 17025 accredited sub-contractor. InnoCal, our internal ISO 17025 accredited laboratory manages this sub-contracted service for our customers ensuring that the strictest quality requirements are being maintained. Calibration reports are issued directly to the customer after incoming inspection is performed by InnoCal's metrology staff. All measurements reported are traceable to SI units.
Specifications
  • DescriptionNIST Calibration Service for Masses. 0.51 to 2 Lb; ASTM 1-3; Oiml F1/F2