Troemner Class-1 Analytical Calibration Weights
ASTM Class 1 - Can be used as a reference standard in calibrating other weights and is appropriate for calibrating high-precision analytical balances with a readability as low as0. 1 mg to 0.01 mg .
  • Surfaces are polished to a perfect, mirror like finish
  • Masses are less vulnerable to wear and scratches
  • Low magnetic susceptibility, uniform consistency and high resistance to corrosion
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity and turned up end for easy handling
  • 1g to 5kg weights are two-piece construction with adjusting cavity and handling knob
Appropriate for calibrating high-precision analytical balances with readability as low as 0.1 mg to 0.01 mg.
The weights are manufactured of the finest stainless steels available, including Troemner Alloy 8 Stainless Steel, which is available exclusively through Troemner. This unique material possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion. Alloy 8 is the finest grade of stainless steel available and is similar to material originally specified in the manufacture of 1 kg state lab primary standards.
Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances
decrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.
Suggested Markets for Analytical Precision Weights include, but are not limited to,Manufacturing QC, Secondary Calibration Labs, State Laboratories, Pharmaceutical QC, R&D, Analytical Testing, Chemical Manufacturing, Utilities.
30,900.00 - ₹676,000.00INR / Each
42 variations of this product are available.
See All Product Options Below
42 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Size (g) Mass Size (mg)
Availability Pricing
Troemner 7037-1W 1 mg, Analytical Class 1 Weight with NVLAP Accredited Certificate 0102122 Mfr # 7037-1W
COMPARE
NVLAP 0.001 1
₹35,000.00INR / Each
Troemner 7036-1W 2 mg, Analytical Class 1 Weight with NVLAP Accredited Certificate 0102123 Mfr # 7036-1W
COMPARE
NVLAP 0.002 2
₹35,000.00INR / Each
Troemner 7034-1W 5 mg, Analytical Class 1 Weight with NVLAP Accredited Certificate 0102124 Mfr # 7034-1W
COMPARE
NVLAP 0.005 5
₹35,000.00INR / Each
Troemner 7032-1W 20 mg, Analytical Class 1 Weight with NVLAP Accredited Certificate 0102125 Mfr # 7032-1W
COMPARE
NVLAP 0.02 20
₹35,000.00INR / Each
Troemner 7030-1W 50 mg, Analytical Class 1 Weight with NVLAP Accredited Certificate 0102126 Mfr # 7030-1W
COMPARE
NVLAP 0.05 50
₹35,000.00INR / Each
Troemner 7029-1W 100 mg, Analytical Class 1 Weight with NVLAP Accredited Certificate 0102127 Mfr # 7029-1W
COMPARE
NVLAP 0.1 100
₹35,000.00INR / Each
Troemner 7028-1W 200 mg, Analytical Class 1 Weight with NVLAP Accredited Certificate 0102128 Mfr # 7028-1W
COMPARE
NVLAP 0.2 200
₹35,000.00INR / Each
Troemner 7026-1W 500 mg, Analytical Class 1 Weight with NVLAP Accredited Certificate 0102129 Mfr # 7026-1W
COMPARE
NVLAP 0.5 500
₹40,800.00INR / Each
Troemner 7013-1W 1kg Analytical Class 1 Weight with NVLAP Accredited Certificate 0102130 Mfr # 7013-1W
COMPARE
NVLAP 1000 1000000
₹194,000.00INR / Each
Troemner 7012-1W 2kg Analytical Class 1 Weight with NVLAP Accredited Certificate 0102131 Mfr # 7012-1W
COMPARE
NVLAP 2000 2000000
₹277,000.00INR / Each
Troemner 7010-1W 5kg Analytical Class 1 Weight with NVLAP Accredited Certificate 0102132 Mfr # 7010-1W
COMPARE
NVLAP 5000 5000000
₹423,000.00INR / Each
Troemner 7009-1W 10kg Analytical Class 1 Weight with NVLAP Accredited Certificate 0102337 Mfr # 7009-1W
COMPARE
NVLAP 10000 100000
₹676,000.00INR / Each
Troemner 7010-1T 5 kg Class 1 Analytical Weight with Traceable Cert 0103127 Mfr # 7010-1T
COMPARE
NIST 5000 5000000
₹381,000.00INR / Each
Troemner 7012-1T 2 kg Class 1 Analytical Weight with Traceable Cert 0103128 Mfr # 7012-1T
COMPARE
NIST 2000 2000000
₹258,000.00INR / Each
Troemner 7013-1T 1 kg Class 1 Analytical Weight with Traceable Cert 0103129 Mfr # 7013-1T
COMPARE
NIST 1000 1000000
₹175,000.00INR / Each
Troemner 7014-1T 500 g Class 1 Analytical Weight with Traceable Cert 0103131 Mfr # 7014-1T
COMPARE
NIST 500 500000
₹85,400.00INR / Each
Troemner 7016-1T 200 g Class 1 Analytical Weight with Traceable Cert 0103132 Mfr # 7016-1T
COMPARE
NIST 200 200000
₹66,500.00INR / Each
Troemner 7017-1T 100 g Class 1 Analytical Weight with Traceable Cert 0103133 Mfr # 7017-1T
COMPARE
NIST 100 100000
₹44,200.00INR / Each
Troemner 7018-1T 50 g Class 1 Analytical Weight with Traceable Cert 0103134 Mfr # 7018-1T
COMPARE
NIST 50 50000
₹42,700.00INR / Each
Troemner 7024-1T 2 g Class 1 Analytical Weight with Traceable Cert 0103139 Mfr # 7024-1T
COMPARE
NIST 2 2000
₹37,200.00INR / Each
Showing 1 - 20 of 42