Imada DI-4B-25 Digital Torque Tester for Air Tools and Impact Wrenches
Representative image only.
Imada DI-4B-25 Digital Torque Tester for Air Tools and Impact Wrenches
Imada | Mfr: DI-4B-25| Item: 2530600
Ideal for pneumatic and impact tool calibration
₹760,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹760,000.00INR / Each
Buy It Together
Current Item
Imada DI-4B-25 Digital Torque Tester for Air Tools and Impact Wrenches
Imada DI-4B-25 Digital Torque Tester for Air Tools and Impact Wrenches
2530600
₹760,000.00INR / Each
+ + =
Total Price 885700.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Heavy duty metal construction
  • Easy to read LED panel displays: torque value in lbf-in, kgf-cm and N-cm (selectable) AND number of blows for selected period (0.1 - 9.9 sec)
  • Peak, Real Time and Peak Down Measuring modes
  • Both CW and CCW operation
  • Accuracy: ±0.5% F.S. ±1 LSD
  • USB output (virtual RS232 serial output)(peak data only)
  • Runs on internal NiCad batteries (8 hour use)
  • Soft and hard joints, AC adapter/charger and Certificate of Calibration traceable to NIST includeD
  • 2 year warranty
More About this Item
lbf-in 3.0~220.0
kgf-cm 3.0~250.0
N-cm 30~2500
Two year warranty
Specifications
  • Accuracy0.5% full scale 1 LSD
  • OutputRS-232
  • DescriptionDigital Torque Tester for Air Tools and Impact Wrenches