Imada SW-1SV-USB Data Acquisition Software for Force and Torque Products
Representative image only.
Imada SW-1SV-USB Data Acquisition Software for Force and Torque Products
Imada | Mfr: SW-1SV-USB| Item: 2530004
Capture key data from your torque testers
₹83,000.00INR / Each
₹83,000.00INR / Each
Product Description
Key Features
  • Customized Excel spreadsheet designed for capturing and analyzing peak and first peak data
  • Ideal for torque testers with either RS-232 or USB ports
  • For Force, Displacement and Torque
  • Full SPC and graphing capabilities
  • Calculates statistical data
More About this Item
Gather peak torque or first peak readings as well as a running log of all torque measurements along with a chart of the data and statistical information. All data is entered directly into a standard Excel spreadsheet, allowing you to analyze your torque tester data using all of the powerful tools available in Excel.
Use SW-1 is a highly customized Excel spreadsheet used to capture and analyze real-time data from Imada force or torque gauges with an RS-232 port. A running log of all the data is displayed along with a chart. Calculate max/min, average, and standard deviation.
Requirements: Imada force or torque gauge with RS-232 port, Pentium or equivalent computer with 32MB RAM or better, a mouse and approximately 1 MB hard disk space, Excel 97 or later.
DS2 REQUIRES CB-203 CABLE
DTX2, HTG2, ZP, ZPH REQUIRE CB-204 CABLE
SW-1 for Windows 95 through Windows XP
SW-1XV for Windows Vista
Specifications
  • DescriptionData Acquisition Software for Force and Torque Products