IKA LR 1000 Basic Reaction Station; 115/230 VAC
Representative image only.
IKA LR 1000 Basic Reaction Station; 115/230 VAC
IKA | Mfr: 0008040101| Item: 2871098
Optimize chemical reactions in your laboratory with these modular reaction systems
₹3,309,000.00INR / Each
₹3,309,000.00INR / Each
Buy It Together
Current Item
IKA LR 1000 Basic Reaction Station; 115/230 VAC
IKA LR 1000 Basic Reaction Station; 115/230 VAC
2871098
₹3,309,000.00INR / Each
+
IKA MVP 10 Basic Diaphragm Vacuum Pump
2871096
₹836,000.00INR / Each
=
Total Price 4145000.00 ADD 2 ITEMS TO CART
Product Description
Includes
Base reactor vessel, temperature sensor, receptacle for temperature sensor, vacuum valve, support rod, 2 pieces of hose connectors, and power cord.
Key Features
  • Digital display for monitoring speed and temperature
  • Included pH monitoring and weighing function (control model only)
  • RS-232 and USB connections for PC control (control model includes an option for Bluetooth)
  • Basic package with fixed display
  • 3 year warranty
More About this Item
Both systems are used to reproduce and enhance chemical reactions as well as to mix, disperse, and homogenize. They have a wide range of uses from general laboratory to pharmaceutical. A digital display allows the user to reach a set temperature and speed in a controlled manner. Ports for output let you control and monitor your reaction right from your PC.
Control models present the addition of pH and weight measurement. Bluetooth interface and separate wireless controller added to increase convenience of use.
3 Year WarrantyIP54 Rated
Specifications
  • Control TypeBasic
  • Width (InchShort)17
  • Height (InchShort)14
  • Max Temperature (° F)248
  • Max Temperature (° C)120
  • Power (VAC)115 / 230
  • Power (watts)1200
  • Combination Block -Tube / Vial Size AcceptedUp to 1000 mL
  • Depth (in)12
  • DescriptionBasic System
  • Warranty3 Years