IKA 2871000 MF 10.2 Impact-Grinding Head for Continuous-Feed Grinding Drive
Representative image only.
IKA 2871000 MF 10.2 Impact-Grinding Head for Continuous-Feed Grinding Drive
IKA | Mfr: 2871000| Item: 0430032
₹513,384.00INR / Each
₹513,384.00INR / Each
Buy It Together
Current Item
IKA 2871000 MF 10.2 Impact-Grinding Head for Continuous-Feed Grinding Drive
IKA 2871000 MF 10.2 Impact-Grinding Head for Continuous-Feed Grinding Drive
0430032
₹513,384.00INR / Each
+ + =
Total Price 626844.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Continuous duty—great for inline applications
  • Surfaces that contact your samples are 304 SS—ideal for sanitary applications
  • 3 month warranty
More About this Item
For use with Continuous-Feed Grinding Drive.
Impact grinding head is ideal for crushing hard brittle materials such a minerals, grains, and pharmaceuticals.
Insert interchangeable sieves (sold separately) into grinding heads to filter maximum particle size.
3 Month Warranty
Specifications
  • Feed Particle Size10 mm max
  • Final Particle Size250 µm
  • DescriptionMF 10.2 Impact-Grinding Head for Continuous-Feed Grinding Drive
  • Warranty3 Months