Idex Sample Loop for PR/EV700-100 & EV750-100, PEEK/Alumina Ceramic, 1 µL, Internal Groove; 1/EA
Representative image only.
Idex Sample Loop for PR/EV700-100 & EV750-100, PEEK/Alumina Ceramic, 1 µL, Internal Groove; 1/EA
Idex | Mfr: 7123-227| Item: 0201675
Biocompatible and inert to almost all organic solvents
₹77,000.00INR / Each
₹77,000.00INR / Each
Buy It Together
Current Item
Idex Sample Loop for PR/EV700-100 & EV750-100, PEEK/Alumina Ceramic, 1 µL, Internal Groove; 1/EA
Idex Sample Loop for PR/EV700-100 & EV750-100, PEEK/Alumina Ceramic, 1 µL, Internal Groove; 1/EA
0201675
₹77,000.00INR / Each
+ =
Total Price 108000.00 ADD 2 ITEMS TO CART
Product Description
Includes
Two unswaged fittings.
Key Features
  • Excellent alternative to stainless steel loops
  • Easy to install or replace
  • 1 µL sample volume
  • 1 year warranty
More About this Item
Biocompatible PEEK polymer sample loops are inert to almost all organic solvents, making them a versatile alternative to stainless steel. The loop ends are cut clean and straight for easy valve installation.
1 Year Warranty
Specifications
  • MaterialPEEK/Alumina Ceramic
  • Compatible WithInjection valve models PR/EV700-100 & EV750-100
  • DescriptionSample Loop for PR/EV700-100 & EV750-100, PEEK/Alumina Ceramic, 1 µL, Internal Groove; 1/EA