Idex Nut/Ferrule Sets, PEEK, for 1/16" OD Sample Loop, 10-32 Coned; 10/Pk
Representative image only.
Idex Nut/Ferrule Sets, PEEK, for 1/16" OD Sample Loop, 10-32 Coned; 10/Pk
Idex | Mfr: 6000-254| Item: 0201674
Two-piece fittings make sample loop replacement a snap
₹31,000.00INR / Pkg of 10
₹31,000.00INR / Pkg of 10
Buy It Together
Current Item
Idex Nut/Ferrule Sets, PEEK, for 1/16" OD Sample Loop, 10-32 Coned; 10/Pk
Idex Nut/Ferrule Sets, PEEK, for 1/16" OD Sample Loop, 10-32 Coned; 10/Pk
0201674
₹31,000.00INR / Pkg of 10
+
Idex Ferrule, PEEK, for 1/16" OD Sample Loop; 10/Pk
0201673
₹15,800.00INR / Pkg of 10
=
Total Price 46800.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • PEEK construction
  • For 1/16" OD tubing
  • 1 year warranty
More About this Item
Choose from nut and ferrule sets or ferrule fittings only to make sample loop replacement quick and easy.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (InchShort)10-32 coned
  • Compression Size / Tube Od (Port 2) (in)1/16
  • ColorNatural
  • DescriptionNut/Ferrule Sets, PEEK, for 1/16" OD Sample Loop, 10-32 Coned; 10/PK