Idex Pressure Relief Valve, Tee Only, 1/16" OD Tubing; 1/EA
Representative image only.
Idex Pressure Relief Valve, Tee Only, 1/16" OD Tubing; 1/EA
Idex | Mfr: P-612| Item: 0201395
Prevent system over pressurization
₹28,000.00INR / Each
₹28,000.00INR / Each
Buy It Together
Current Item
Idex Pressure Relief Valve, Tee Only, 1/16" OD Tubing; 1/EA
Idex Pressure Relief Valve, Tee Only, 1/16" OD Tubing; 1/EA
0201395
₹28,000.00INR / Each
+ + =
Total Price 73000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 0
  • 14 µL swept volume
  • 1 month warranty
More About this Item
Relief valves protect system components by diverting fluid automatically when inline pressure exceeds the set limit. Use the 100 psi model for general-purpose applications; 5 psi model is perfect for protecting syringe and peristaltic pump systems. To have the pressure relief valve open at a different pressure than 5 or 100 psi, combine a pressure relief tee assembly with a back pressure regulator assembly.
1 Month Warranty
Specifications
  • Connection TypeHPLC/Chromatography
  • Housing MaterialPCTFE
  • Tubing OD (in)0.0625
  • DescriptionIdex P-612 Pressure Relief Valve, Tee Only, 1/16" OD Tubing; 1/EA