Idex Low-Pressure Union, Male Straight, PEEK, 0.062" Bore, 1/4-28 Flat Bottom; 1/EA
Representative image only.
Idex Low-Pressure Union, Male Straight, PEEK, 0.062" Bore, 1/4-28 Flat Bottom; 1/EA
Idex | Mfr: P-645| Item: 0201407
Make solid connections in low-pressure applications
₹4,900.00INR / Each
₹4,900.00INR / Each
Buy It Together
Current Item
Idex Low-Pressure Union, Male Straight, PEEK, 0.062" Bore, 1/4-28 Flat Bottom; 1/EA
Idex Low-Pressure Union, Male Straight, PEEK, 0.062" Bore, 1/4-28 Flat Bottom; 1/EA
0201407
₹4,900.00INR / Each
+ =
Total Price 6400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Manufactured from inert polymers
  • Provide your own fittings to suit your application
More About this Item
Use straight unions in a variety of low-pressure situations throughout your fluidic system. Use straight and elbow unions in a variety of low-pressure situations throughout your fluidic system. Unions do not include fittings, allowing for customization of fittings.
1 Month Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (InchShort)1/4-28 Flat Bottom
  • ColorNatural
  • DescriptionLow-Pressure Union, Male Straight, Natural PCTFE, 0.062" Bore, 1/4-28 Flat Bottom; 1/EA