IDEX® Stainless Steel Tubing, 1/16" OD
Precut stainless steel tubing ready for all chromatography testing requirements
  • 316 stainless steel tubing, polished and passivated
  • Color coded for easy identification
  • Suited for most analytical applications
IDEX 316 stainless steel tubing is precut and thoroughly passivated (except lengths 25 feet and beyond), making it the recommended choice for most chromatographic analytical applications. Tubing is machine cut, polished. and each end is deburred inside and out. Then it is submerged in an acid passivation bath and flushed clean. Each tube is color coded for easy identification of inner diameter.
1,500.00 - ₹504,000.00INR / Each
62 variations of this product are available.
See All Product Options Below
62 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Pressure (PSI) Tubing ID (in)
Availability Pricing
Idex U-101 Chromatography Tubing, 316 SS, 1/16" OD x 0.020" ID x 2" L, Yellow; 1/EA 0201082 Mfr # U-101
COMPARE
0.167 15800 1/50
₹1,500.00INR / Each
Idex U-103 Chromatography Tubing, 316 SS, 1/16" OD x 0.020" ID x 8" L, Yellow; 1/EA 0201084 Mfr # U-103
COMPARE
0.666 15800 1/50
₹2,600.00INR / Each
Idex U-104 Chromatography Tubing, 316 SS, 1/16" OD x 0.020" ID x 12" L, Yellow; 1/EA 0201085 Mfr # U-104
COMPARE
1 15800 1/50
₹3,200.00INR / Each
Idex U-106 Chromatography Tubing, 316 SS, 1/16" OD x 0.010" ID x 5 ft L, Blue Band; 1/EA 0201087 Mfr # U-106
COMPARE
5 19700 1/100
₹13,400.00INR / Each
Idex U-107 Chromatography Tubing, 316 SS, 1/16" OD x 0.030" ID x 5 ft L; 1/EA  0201088 Mfr # U-107
COMPARE
5 12000 1/32
₹13,200.00INR / Each
Idex U-108 Chromatography Tubing, 316 SS, 1/16" OD x 0.007" ID x 5 ft L, Black; 1/EA 0201089 Mfr # U-108
COMPARE
5 20900 1/150
₹14,000.00INR / Each
Idex U-111 Chromatography Tubing, 316 SS, 1/16" OD x .010" ID x 5 cm L; 1/EA  0201090 Mfr # U-111
COMPARE
0.167 19700 1/100
₹1,600.00INR / Each
Idex U-112 Chromatography Tubing, 316 SS, 1/16" OD x 0.010" ID x 10 cm L; 1/EA  0201096 Mfr # U-112
COMPARE
0.333 19700 1/100
₹1,900.00INR / Each
Idex Chromatography Tubing, 316 Stainless Steel, 1/16" OD x 0.010" ID x 8" L, Blue 0201105 Mfr # U-113
COMPARE
0.666 19700 1/100
₹2,600.00INR / Each
Idex U-114 Chromatography Tubing, 316 SS, 1/16" OD x 0.010" ID x 30 cm L; 1/EA  0201112 Mfr # U-114
COMPARE
1 19700 1/100
₹3,800.00INR / Each
Idex U-115 Chromatography Tubing, 316 SS, 1/16" OD x 0.030" ID x 2" L, White; 1/EA 0201120 Mfr # U-115
COMPARE
0.167 12000 1/32
₹1,600.00INR / Each
Idex U-116 Chromatography Tubing, 316 SS, 1/16" OD x 0.030" ID x 4" L, White; 1/EA 0201121 Mfr # U-116
COMPARE
0.333 12000 1/32
₹2,400.00INR / Each
Idex U-117 Chromatography Tubing, 316 SS, 1/16" OD x 0.030" ID x 8" L, White; 1/EA 0201122 Mfr # U-117
COMPARE
0.666 12000 1/32
₹2,900.00INR / Each
Idex U-118 Chromatography Tubing, 316 SS, 1/16" OD x 0.030" ID x 12" L, White; 1/EA 0201123 Mfr # U-118
COMPARE
1 12000 1/32
₹3,900.00INR / Each
Idex U-126 Chromatography Tubing, 316 SS, 1/16" OD x 0.007" ID x 2" L, Black; 1/EA 0201124 Mfr # U-126
COMPARE
0.167 20900 1/150
₹1,700.00INR / Each
Idex U-127 Chromatography Tubing, 316 SS, 1/16" OD x 0.007" ID x 4" L, Black; 1/EA 0201125 Mfr # U-127
COMPARE
0.333 20900 1/150
₹2,000.00INR / Each
Idex U-128 Chromatography Tubing, 316 SS, 1/16" OD x 0.007" ID x 8" L, Black; 1/EA 0201126 Mfr # U-128
COMPARE
0.666 20900 1/150
₹2,800.00INR / Each
Idex U-129 Chromatography Tubing, 316 SS, 1/16" OD x 0.007" ID x 12" L, Black; 1/EA 0201127 Mfr # U-129
COMPARE
1 20900 1/150
₹10,400.00INR / Each
Idex U-130 Chromatography Tubing, 316 SS, 1/16" OD x 0.007" ID x 1.6 ft L, Black; 1/EA 0201128 Mfr # U-130
COMPARE
1.6 20900 1/150
₹5,700.00INR / Each
Idex U-144 Chromatography Tubing, 316 SS, 1/16" OD x 0.040" ID x 5 ft L, Natural; 1/EA 0201142 Mfr # U-144
COMPARE
5 8100 1/25
₹33,000.00INR / Each
Showing 1 - 20 of 62