Idex Low-Pressure Union, Female Straight, Natural ETFE, 0.030" Bore, 1/4-28 Flat Bottom; 1/EA
Representative image only.
Idex Low-Pressure Union, Female Straight, Natural ETFE, 0.030" Bore, 1/4-28 Flat Bottom; 1/EA
Idex | Mfr: P-710-01| Item: 0202373
Make solid connections in low-pressure applications
₹4,100.00INR / Each
₹4,100.00INR / Each
Buy It Together
Current Item
Idex Low-Pressure Union, Female Straight, Natural ETFE, 0.030" Bore, 1/4-28 Flat Bottom; 1/EA
Idex Low-Pressure Union, Female Straight, Natural ETFE, 0.030" Bore, 1/4-28 Flat Bottom; 1/EA
0202373
₹4,100.00INR / Each
+
Idex Pressure Relief Valve, Tee Only, 1/16" OD Tubing; 1/EA
0201395
₹28,000.00INR / Each
+ + + + =
Total Price 52630.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Unions do not include fittings—provide your own to match your tubing size
  • Manufactured from inert polymers
  • 0.030" thru-hole diameter
  • ETFE body
  • 1 year warranty
More About this Item
Use straight and elbow unions in a variety of low-pressure situations throughout your fluidic system. Unions do not include fittings, allowing for customization of fittings. Connect two different size OD tubing by selecting flangeless fittings for 1/16" OD tubing and 1/8" OD tubing.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(F) (port 1) (InchShort)1/4-28 Flat Bottom
  • ColorNatural
  • DescriptionLow-Pressure Union, Female Straight, Natural ETFE, 0.030" Bore, 1/4-28 Flat Bottom; 1/EA