Idex Low-Pressure Union, Female Straight, Black Acetal, 0.020" Bore, M6 Flat Bottom; 1/EA
Representative image only.
Idex Low-Pressure Union, Female Straight, Black Acetal, 0.020" Bore, M6 Flat Bottom; 1/EA
Idex | Mfr: P-602| Item: 0202336
Make solid connections in low-pressure applications
₹1,800.00INR / Each
₹1,800.00INR / Each
Buy It Together
Current Item
Idex Low-Pressure Union, Female Straight, Black Acetal, 0.020" Bore, M6 Flat Bottom; 1/EA
Idex Low-Pressure Union, Female Straight, Black Acetal, 0.020" Bore, M6 Flat Bottom; 1/EA
0202336
₹1,800.00INR / Each
+ =
Total Price 3300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Manufactured from inert polymers
  • Provide your own fittings to suit your application
  • M6 flat-bottom ports
  • 0.020" thru-hole diameter
  • Black acetal body
  • 1 year warranty
More About this Item
Use straight unions in a variety of low-pressure situations throughout your fluidic system. Use straight and elbow unions in a variety of low-pressure situations throughout your fluidic system. Unions do not include fittings, allowing for customization of fittings.
1 Year Warranty
Specifications
  • Fitting MaterialAcetal
  • Connection TypeHPLC/Chromatography
  • Thread Size ISO Metric (F) (Port 1) (mm)6
  • ColorBlack
  • DescriptionLow-Pressure Union, Female Straight, Black Acetal, 0.020" Bore, M6 Flat Bottom; 1/EA