Idex Inlet Solvent Filter, Stem, SS, PCTFE Flangeless Fitting, 10 µm, 1/8" OD Tubing; 1/EA
Representative image only.
Idex Inlet Solvent Filter, Stem, SS, PCTFE Flangeless Fitting, 10 µm, 1/8" OD Tubing; 1/EA
Idex | Mfr: A-302A| Item: 4270102
Keep damaging particles out of your system with inlet solvent filters
₹13,900.00INR / Each
₹13,900.00INR / Each
Buy It Together
Current Item
Idex Inlet Solvent Filter, Stem, SS, PCTFE Flangeless Fitting, 10 µm, 1/8" OD Tubing; 1/EA
Idex Inlet Solvent Filter, Stem, SS, PCTFE Flangeless Fitting, 10 µm, 1/8" OD Tubing; 1/EA
4270102
₹13,900.00INR / Each
+ + =
Total Price 19550.00 ADD 3 ITEMS TO CART
Product Description
Includes
One XP-325 PCTFE flangeless fitting
Key Features
  • Protect your system from particulate matter that can cause damage
  • Filters available to fit specific system specifications
  • Easy to replace without tools
More About this Item
For good preventive maintenance, inlet solvent filters are an optimal way to cleanse solvents, avoiding pump cavitation and system damage. Different style stainless steel filters are available for specific system specifications and can be easily replaced regularly without tools.
To be used with 1/8" OD tubing.
Specifications
  • Membrane MaterialStainless Steel
  • Housing MaterialPCTFE
  • Pore Size (µm)10
  • Connections1/4-28 flat bottom
  • DescriptionInlet Solvent Filter, Stem, SS, PCTFE Flangeless Fitting, 10 µm, 1/8" OD Tubing; 1/EA