Idex Flangeless Male Nut, Short Head, Yellow ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Representative image only.
Idex Flangeless Male Nut, Short Head, Yellow ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Idex | Mfr: P-318Y| Item: 0202009
Excellent for tight-space applications
₹1,700.00INR / Pkg of 1
₹1,700.00INR / Pkg of 1
Buy It Together
Current Item
Idex Flangeless Male Nut, Short Head, Yellow ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Idex Flangeless Male Nut, Short Head, Yellow ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
0202009
₹1,700.00INR / Pkg of 1
+
Idex Lock Nut for 1/4-28 Flangeless Fittings, White Acetal; 10/PK
0201997
₹9,800.00INR / Pkg of 10
+ +
Cole-Parmer FEP Tubing, 1/16" ID x 1/8" OD; 50 Ft
5011915
₹13,300.00INR / Pkg of 1
=
Total Price 29400.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Yellow ETFE
  • 10/PK
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorYellow
  • DescriptionFlangeless Male Nut, Short Head, Yellow ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK