Cole-Parmer Sterilizing Air Filter
Sterile filter with hydrophobic PTFE membrane for degassing bioreactors
  • Stepped hose barb connections accept 1/4" to 3/8" inner diameter tubing
This venting disc filter has a hydrophobic PTFE membrane and PP housing—making it ideal for venting and degassing fermentation tanks and culture vessels, drying or sterilizing gases, and cleaning analytical solvents and reagents. Filter can withstand up to 30 autoclave cycles at 121°C for 20 minutes or 3 cycles at 134°C for 30 minutes.
550.00 - ₹4,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Membrane Material Pore Size (µm)
Availability Pricing
COMPARE
50 Polytetrafluoroethylene (PTFE) 0.2
₹4,300.00INR / Each
COMPARE
50 Polytetrafluoroethylene (PTFE) 0.2
₹550.00INR / Each