Idex Flangeless Male Nut, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Male Nut, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Idex | Mfr: P-335| Item: 0202023
Excellent for tight-space applications
₹1,200.00INR / Each
₹1,200.00INR / Each
Buy It Together
Current Item
Idex Flangeless Male Nut, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Idex Flangeless Male Nut, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
0202023
₹1,200.00INR / Each
+ + +
Cole-Parmer FEP Tubing, 1/16" ID x 1/8" OD; 50 Ft
5011915
₹13,300.00INR / Pkg of 1
=
Total Price 20200.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Natural PEEK
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorNatural
  • DescriptionFlangeless Male Nut, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA