Idex Flangeless Male Nut, Large Knurl, Black PEEK, 1/4" OD Tubing, 1/2-20 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Male Nut, Large Knurl, Black PEEK, 1/4" OD Tubing, 1/2-20 Flat-Bottom; 1/EA
Idex | Mfr: U-655| Item: 0202049
Eliminate the need to flange tubing
₹2,300.00INR / Each
₹2,300.00INR / Each
Buy It Together
Current Item
Idex Flangeless Male Nut, Large Knurl, Black PEEK, 1/4" OD Tubing, 1/2-20 Flat-Bottom; 1/EA
Idex Flangeless Male Nut, Large Knurl, Black PEEK, 1/4" OD Tubing, 1/2-20 Flat-Bottom; 1/EA
0202049
₹2,300.00INR / Each
+ =
Total Price 3010.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Removable and reusable—saving you money!
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/4
  • ColorBlack