Idex Bottom-of-the-Bottle™ Inlet Solvent Filter Assembly, SS/PEEK/ETFE, 10 µm, 1/8" OD Tubing; 1/EA
Representative image only.
Idex Bottom-of-the-Bottle™ Inlet Solvent Filter Assembly, SS/PEEK/ETFE, 10 µm, 1/8" OD Tubing; 1/EA
Idex | Mfr: A-550| Item: 4270002
Protect your system from damaging particulates
₹38,200.00INR / Each
₹38,200.00INR / Each
Buy It Together
Current Item
Idex Bottom-of-the-Bottle™ Inlet Solvent Filter Assembly, SS/PEEK/ETFE, 10 µm, 1/8" OD Tubing; 1/EA
Idex Bottom-of-the-Bottle™ Inlet Solvent Filter Assembly, SS/PEEK/ETFE, 10 µm, 1/8" OD Tubing; 1/EA
4270002
₹38,200.00INR / Each
+ + =
Total Price 71700.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
One 42700-52 SS filter cup and one flangeless fitting.
Key Features
  • Economical—stainless steel filter cups are replaceable
  • Max flow rate of 40 mL/min
More About this Item
Stainless steel bottom-of-the-bottle filters allow solvent to be drawn from within 1/8” of the bottom of your solvent bottle. Filters are constructed of PEEK, ETFE, and SS.
Specifications
  • Membrane MaterialPEEK
  • Pore Size (micron)10
  • Housing MaterialStainless steel
  • Connections1/8" OD tubing
  • Typical Water Flow Rate40 mL/min (max)
  • DescriptionBottom-of-the-Bottle™ Inlet Solvent Filter Assembly, SS/PEEK/ETFE, 10 µm, 1/8" OD Tubing; 1/EA