Idex Bottom-of-the-Bottle™ Replacement Filter Cup, SS, 10 µm; 10/PK

Representative image only.

Idex Bottom-of-the-Bottle™ Replacement Filter Cup, SS, 10 µm; 10/PK

Idex | Mfr: A-520X| Item: 4270052

Save money and time using filter cups

₹28,100.00INR / Pkg of 10

₹28,100.00INR / Pkg of 10
Product Description

Key Features

  • Optimize performance of the Idex Bottom-of-the-Bottle™ inlet solvent filter assemblies with these replacement cups
  • Max flow rate 40 mL/min

More About this Item

The replaceable filter cups are inexpensive and easy to replace.

Specifications
  • MaterialStainless steel and PEEK
  • Compatible WithFilter 42700-02
  • DescriptionBottom-of-the-Bottle™ Replacement Filter Cup, SS, 10 µm; 10/PK
Reviews
Q&A