Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, UHMWPF/ETFE, 10 µm, 1/16" OD Tubing; 1/EA
Representative image only.
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, UHMWPF/ETFE, 10 µm, 1/16" OD Tubing; 1/EA
Idex | Mfr: A-445| Item: 4271146
Protect your system from damaging particulates
₹10,400.00INR / Each
₹10,400.00INR / Each
Buy It Together
Current Item
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, UHMWPF/ETFE, 10 µm, 1/16" OD Tubing; 1/EA
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, UHMWPF/ETFE, 10 µm, 1/16" OD Tubing; 1/EA
4271146
₹10,400.00INR / Each
+ =
Total Price 29000.00 ADD 2 ITEMS TO CART
Product Description
Includes
One flangeless fitting
Key Features
  • Filters are biocompatible
  • Economical—filter cups are replaceable
  • Use with 1/16" OD tubing
  • 1 year warranty
More About this Item
UHMWPE Filters allow tubing to pass through the bottom of the filter cup to allow solvent to be drawn within 0.10” (2.5 mm) of the bottom of your solvent bottle. Filters are constructed of UHMWPE and ETFE.
UHMWPE is a hydrophobic material. You may need to prime the filter with methanol or acetonitrile to establish proper surface wetting.
1 Year Warranty
Specifications
  • Membrane MaterialUHMWPE
  • Pore Size (micron)10
  • Housing MaterialUHMWPE
  • Connections1/16" OD tubing
  • Typical Water Flow Rate5 mL/min (max)
  • DescriptionBottom-of-the-Bottle™ Biocompatible Filter Assembly, UHMWPF/ETFE, 10 µm, 1/16" OD Tubing; 1/EA