Idex Bottom-of-the-Bottle™ Biocompatible Replacement Filter Cup, UHMWPE, 10 µm; 5/PK
Representative image only.
Idex Bottom-of-the-Bottle™ Biocompatible Replacement Filter Cup, UHMWPE, 10 µm; 5/PK
Idex | Mfr: A-427| Item: 4271139
Save money and time using filter cups
₹18,600.00INR / Pkg of 5
₹18,600.00INR / Pkg of 5
Product Description
Key Features
  • Optimize performance of the Idex Bottom-of-the-Bottle™ inlet solvent filter assemblies with these replacement cups
  • Max flow rate 5 mL/min
  • 1 year warranty
More About this Item
The replaceable filter cups are inexpensive and easy to replace.
1 Year Warranty
Specifications
  • MaterialUHMWPE
  • Compatible WithFilters 42711-46 and -47
  • DescriptionBottom-of-the-Bottle™ Biocompatible Replacement Filter Cup, UHMWPE, 10 µm; 5/PK