Idex Barbed Luer Adapter, Polypropylene, 0.080" Bore, Female Luer Lock x 1/8" Hose Barb; 1/EA
Representative image only.
Idex Barbed Luer Adapter, Polypropylene, 0.080" Bore, Female Luer Lock x 1/8" Hose Barb; 1/EA
Idex | Mfr: P-859| Item: 0202406
Excellent for connecting soft-wall tubing to a syringe or a low-pressure filter with luer connection
₹340.00INR / Each
₹340.00INR / Each
Product Description
Key Features
  • Choose from polypropylene or ETFE construction
  • Female Luer Lock x 1/8" Hose Barb
  • Manufactured from polypropylene
  • 1 year warranty
More About this Item
Female luer fittings come with a lock. Male luer fittings are available with slip or lock connection.
1 Year Warranty
Specifications
  • MaterialPolypropylene
  • Connection Type-Port 1Female luer lock
  • Barb Size-Port 2 (in)1/8
  • DescriptionLow-Pressure Adapter, Barbed, Polypropylene, 0.080" Bore, Female Luer Lock x 1/8" Hose Barb; 1/EA