Value Plastics Male Luer with Integral Lock Ring to 200 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
  • Rotating locks minimize twisting and kinking of tubing during positioning
  • Tapered cone seals are reliable and uncomplicated
  • Facilitate single-use configurations that minimize downtime
These luer fittings come in many configurations and several materials, joining tubing from 1/16” (1.6 mm) ID to 1/4” (6.4 mm) ID. The integral lock ring is a built-in mechanism that provides a secure connection to the female luer to prevent accidental disconnection and stop leakage or contamination. The 200 series barb offers outstanding performance characteristics when used with flexible tubing. Tubing is expanded by 50% above its nominal inside diameter, permitting high-pressure capabilities. Because the fittings are made to ISO standards that are well understood globally, it ensures proper interconnections and often removes the need to retrofit*.
Options include nylon, polycarbonate, or polypropylene resin.
Common features of nylon include a high mechanical strength, great balance of stiffness and toughness, good performance at high temperatures, good electrical and flammability properties, and resistance against abrasion or chemicals. It can be sterilized using gamma radiation, Ethylene oxide (EtO), or autoclave.
Polypropylene is clarified and produced without any animal derived components or phthalates. It can be sterilized using gamma radiation, Ethylene oxide (EtO), E-beam, or autoclave.
Polycarbonate is transparent and features high impact, chemical, UV, and electrical resistance. Sterilization is only recommended via ethylene oxide (EtO). Gamma radiation up to 10 Mrads is possible with minor loss of physical properties – it will discolor to a yellow-green hue. Autoclave is not recommended and may crack the fitting.
2,700.00 - ₹3,900.00INR / Pkg of 25
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring x 200 Series Barb 1/16" ID; 25/Pk 5012021 Mfr # MTLL210-1
COMPARE
1/16 Nylon
₹2,700.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Clear Polycarbonate, Male Luer Integral Lock Ring x 200 Series Barb 1/16" ID; 25/Pk 5012022 Mfr # MTLL210-9
COMPARE
1/16 Polycarbonate
₹3,200.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring x 200 Series Barb 3/32" ID; 25/Pk 5012023 Mfr # MTLL220-1
COMPARE
3/32 Nylon
₹2,700.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer Integral Lock Ring x 200 Series Barb 3/32" ID; 25/Pk 5012024 Mfr # MTLL220-6005
COMPARE
3/32 Polypropylene
₹3,400.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Clear Polycarbonate, Male Luer Integral Lock Ring x 200 Series Barb 3/32" ID; 25/Pk 5012025 Mfr # MTLL220-9
COMPARE
3/32 Polycarbonate
₹3,200.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring x 200 Series Barb 1/8" ID; 25/Pk 5012026 Mfr # MTLL230-1
COMPARE
1/8 Nylon
₹2,700.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Clear Polycarbonate, Male Luer Integral Lock Ring x 200 Series Barb 1/8" ID; 25/Pk 5012027 Mfr # MTLL230-9
COMPARE
1/8 Polycarbonate
₹3,200.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring x 200 Series Barb 5/32" ID; 25/Pk 5012028 Mfr # MTLL240-1
COMPARE
5/32 Nylon
₹3,100.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer Integral Lock Ring x 200 Series Barb 5/32" ID; 25/Pk 5012029 Mfr # MTLL240-6005
COMPARE
5/32 Polypropylene
₹3,900.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Clear Polycarbonate, Male Luer Integral Lock Ring x 200 Series Barb 5/32" ID; 25/Pk 5012030 Mfr # MTLL240-9
COMPARE
5/32 Polycarbonate
₹3,500.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer Integral Lock Ring x 200 Series Barb 3/16" ID; 25/Pk 5012031 Mfr # MTLL250-6005
COMPARE
3/16 Polypropylene
₹3,400.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Clear Polycarbonate, Male Luer Integral Lock Ring x 200 Series Barb 3/16" ID; 25/Pk 5012032 Mfr # MTLL250-9
COMPARE
3/16 Polycarbonate
₹3,500.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer with Integral Lock Ring x 200 Series Barb 1/16"; 25/Pk 5012095 Mfr # MTLL210-6005
COMPARE
1/16 Polypropylene
₹3,900.00INR / Pkg of 25
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer with Integral Lock Ring x 200 Series Barb 1/8"; 25/Pk 5012096 Mfr # MTLL230-6005
COMPARE
1/8 Polypropylene
₹2,800.00INR / Pkg of 25
Showing 1- 14 of 14