Idex Barbed Luer Adapter, Polypropylene, 0.069" Bore, Male Slip Luer x 3/32" Hose Barb; 1/EA
Representative image only.
Idex Barbed Luer Adapter, Polypropylene, 0.069" Bore, Male Slip Luer x 3/32" Hose Barb; 1/EA
Idex | Mfr: P-855| Item: 0202403
Excellent for connecting soft-wall tubing to a syringe or a low-pressure filter with luer connection
₹330.00INR / Each
₹330.00INR / Each
Product Description
Key Features
  • Choose from polypropylene or ETFE construction
  • Male Slip Luer x 3/32" Hose Barb
  • 1 year warranty
More About this Item
Female luer fittings come with a lock. Male luer fittings are available with slip or lock connection.
For use in systemsmfor which luer lock rings are not desired.
1 Year Warranty
Specifications
  • MaterialPolypropylene
  • Connection Type-Port 1Male slip luer
  • Barb Size-Port 2 (in)3/32
  • DescriptionLow-Pressure Adapter, Barbed, Polypropylene, 0.069" Bore, Male Slip Luer x 3/32" Hose Barb; 1/EA