Idex Barbed Luer Adapter, Polypropylene, 0.049" Bore, Male Luer Lock x 1/16" Hose Barb; 1/EA
Representative image only.
Idex Barbed Luer Adapter, Polypropylene, 0.049" Bore, Male Luer Lock x 1/16" Hose Barb; 1/EA
Idex | Mfr: P-850| Item: 0202398
Excellent for connecting soft-wall tubing to a syringe or a low-pressure filter with luer connection
₹340.00INR / Each
₹340.00INR / Each
Product Description
Key Features
  • Choose from polypropylene or ETFE construction
  • Male Luer Lock x 1/16" Hose Barb
  • 1 year warranty
More About this Item
Female luer fittings come with a lock. Male luer fittings are available with slip or lock connection.
1 Year Warranty
Specifications
  • MaterialPolypropylene
  • Connection Type-Port 1Male luer lock
  • Barb Size-Port 2 (in)1/16
  • DescriptionLow-Pressure Adapter, Barbed, Polypropylene, 0.049" Bore, Male Luer Lock x 1/16" Hose Barb; 1/EA