Idex 316 Stainless Steel Nut, Valco/VICI Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK
Representative image only.
Idex 316 Stainless Steel Nut, Valco/VICI Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK
Idex | Mfr: U-320X| Item: 0201468
Stainless steel fittings withstand pressures up to 20,000 psi (1380 bar)!
₹15,100.00INR / Pkg of 10
₹15,100.00INR / Pkg of 10
Buy It Together
Current Item
Idex 316 Stainless Steel Nut, Valco/VICI Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK
Idex 316 Stainless Steel Nut, Valco/VICI Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK
0201468
₹15,100.00INR / Pkg of 10
+ =
Total Price 16300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Constructed of durable stainless steel
  • Valco/VICI Compatible
  • 10-32 coned configuration
  • 10/PK
  • 1 month warranty
  • 1 month warranty
More About this Item
These 316 stainless steel nuts (and ferrules) are rated to 20,000 psi (1380 bar) when tightened using a wrench. Recommended torque for tightening these fittings is 20 in-lbs. (2.25 N-m).
Metal fittings should not be used with plastic ports because they can damage the ports.
1 Month Warranty
Specifications
  • Fitting MaterialStainless steel
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (InchShort)10-32 coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.588
  • Max Operating Pressure (PSI)20000
  • Description316 Stainless Steel Nut, Valco/VICI Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK