Idex 316 Stainless Steel Ferrule, Valco/VICI Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK
Representative image only.
Idex 316 Stainless Steel Ferrule, Valco/VICI Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK
Idex | Mfr: U-321X| Item: 0201469
Can withstand pressures up to 20,000 psi (1380 bar)!
₹10,100.00INR / Pkg of 10
₹10,100.00INR / Pkg of 10
Buy It Together
Current Item
Idex 316 Stainless Steel Ferrule, Valco/VICI Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK
Idex 316 Stainless Steel Ferrule, Valco/VICI Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK
0201469
₹10,100.00INR / Pkg of 10
+ =
Total Price 11600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed of durable stainless steel
  • Waters Compatible
  • 10-32 coned configuration
  • 10/PK
  • 1 month warranty
  • 1 month warranty
More About this Item
Use with IDEX high-pressure stainless steel nuts for your high-pressure applications. Stainless steel ferrules are rated to 20,000 psi (1380 bar) when tightened using a wrench. Recommended torque for tightening these fittings is 20 in-lbs. (2.25 N-m).
Metal fittings should not be used with plastic ports because they can damage the ports.
1 Month Warranty
Specifications
  • Material316 stainless steel
  • Compatible WithNut 02014-68
  • Description316 Stainless Steel Ferrule, Valco/VICI Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK